Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CRY1 SIRNA Assays ›
          Gene Symbol:
          Cry1
          Gene Name:
          cryptochrome 1 (photolyase-like)
          Gene Aliases:
          AU020726, AU021000, Phll1
          Chromosome Location:
          Chr.10: 85131700 - 85185054 on Build GRCm38
          UniGene ID:
          Mm.26237
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS203274
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cry1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007771.3 NP_031797.1 10 2099
          GenBank mRNA AB000777.1 - 10 1535
          AF156986.1 - 10 1834
          AK133818.1 - 10 2111
          AK162460.1 - 10 2099
          BC022174.1 - 10 1870 4036758
          BC085499.1 - 10 2099 6813367

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA photolyase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          gluconeogenesis
          DNA repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA damage induced protein phosphorylation
          circadian rhythm
          protein-chromophore linkage
          lipid storage
          negative regulation of protein ubiquitination
          response to insulin
          circadian regulation of gene expression
          response to glucagon
          glucose homeostasis
          regulation of circadian rhythm
          negative regulation of circadian rhythm
          entrainment of circadian clock by photoperiod
          negative regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of transcription, DNA-templated
          rhythmic process
          response to stimulus
          regulation of DNA damage checkpoint
          negative regulation of glucocorticoid receptor signaling pathway
          negative regulation of glucocorticoid secretion
          nucleotide binding
          transcription factor activity, transcription factor binding
          core promoter sequence-specific DNA binding
          core promoter binding
          double-stranded DNA binding
          DNA photolyase activity
          protein binding
          transcription factor binding
          photoreceptor activity
          kinase binding
          protein kinase binding
          phosphatase binding
          nuclear hormone receptor binding
          histone deacetylase binding
          ubiquitin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nbkcc:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0