Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010020.3 | NP_034150.1 | 7, 8 | 1180 | ||
GenBank mRNA | AF109072.1 | - | 7, 8 | 1103 | ||
AF109391.1 | - | 6, 7 | 1028 | |||
AJ238309.1 | - | 6, 7 | 1036 | |||
AK032629.1 | - | 7, 8 | 1138 | |||
AK136885.1 | - | 7, 8 | 1125 | |||
BC054119.1 | - | 7, 8 | 1180 | 6588897 |