Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DRD2 SIRNA Assays ›
          Gene Symbol:
          Drd2
          Gene Name:
          dopamine receptor D2
          Gene Aliases:
          D2R, Drd-2
          Chromosome Location:
          Chr.9: 49340428 - 49408174 on Build GRCm38
          UniGene ID:
          Mm.41970
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS203672
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Drd2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010077.2 NP_034207.2 6 877
          GenBank mRNA BC105663.1 - 6 877 40046665
          BC105664.1 - 6 877 40046668
          BC105665.1 - 6 877 40046660
          BC105666.1 - 6 877 40046662
          X55674.1 - 6 912

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_000795, AB209832, X51645, AK308777, BC021195, and others.
          This siRNA also targets the following transcripts in Rat: NM_012547, XM_008766190, X56065, M32241, X53278.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          response to hypoxia
          synaptic transmission, dopaminergic
          startle response
          response to amphetamine
          neurological system process involved in regulation of systemic arterial blood pressure
          regulation of heart rate
          regulation of sodium ion transport
          G-protein coupled receptor internalization
          positive regulation of neuroblast proliferation
          positive regulation of receptor internalization
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          dopamine receptor signaling pathway
          chemical synaptic transmission
          neuron-neuron synaptic transmission
          axonogenesis
          sensory perception of smell
          long-term memory
          grooming behavior
          locomotory behavior
          adult walking behavior
          feeding behavior
          protein localization
          negative regulation of cell proliferation
          associative learning
          visual learning
          response to light stimulus
          response to toxic substance
          regulation of dopamine secretion
          Wnt signaling pathway
          cerebral cortex GABAergic interneuron migration
          adenohypophysis development
          negative regulation of cell migration
          peristalsis
          adult behavior
          regulation of cAMP metabolic process
          forebrain development
          auditory behavior
          activation of protein kinase activity
          regulation of synaptic transmission, GABAergic
          positive regulation of cytokinesis
          circadian regulation of gene expression
          negative regulation of dopamine secretion
          response to histamine
          intracellular signal transduction
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          positive regulation of multicellular organism growth
          response to cocaine
          negative regulation of circadian sleep/wake cycle, sleep
          dopamine metabolic process
          response to drug
          regulation of potassium ion transport
          response to morphine
          regulation of MAPK cascade
          pigmentation
          regulation of phosphoprotein phosphatase activity
          positive regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of blood pressure
          positive regulation of transcription from RNA polymerase II promoter
          phosphatidylinositol metabolic process
          negative regulation of insulin secretion
          acid secretion
          behavioral response to cocaine
          behavioral response to ethanol
          regulation of long-term neuronal synaptic plasticity
          branching morphogenesis of a nerve
          arachidonic acid secretion
          negative regulation of protein secretion
          modulation of synaptic transmission
          release of sequestered calcium ion into cytosol
          negative regulation of cytosolic calcium ion concentration
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          regulation of dopamine uptake involved in synaptic transmission
          positive regulation of dopamine uptake involved in synaptic transmission
          regulation of synapse structural plasticity
          negative regulation of protein kinase B signaling
          negative regulation of synaptic transmission, glutamatergic
          positive regulation of growth hormone secretion
          prepulse inhibition
          phospholipase C-activating dopamine receptor signaling pathway
          negative regulation of dopamine receptor signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          regulation of locomotion involved in locomotory behavior
          positive regulation of glial cell-derived neurotrophic factor secretion
          positive regulation of long-term synaptic potentiation
          negative regulation of voltage-gated calcium channel activity
          dopamine neurotransmitter receptor activity, coupled via Gi/Go
          signal transducer activity
          G-protein coupled receptor activity
          dopamine neurotransmitter receptor activity
          receptor binding
          drug binding
          dopamine binding
          ionotropic glutamate receptor binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mtlvn:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0