Search
Search

Epb4.1l3| Gene Symbol: Epb4.1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013813.1 | NP_038841.1 | 16, 17 | 2101 | ||
| GenBank mRNA | AB370264.1 | - | 15, 16 | 1989 | ||
| AB523362.1 | - | 15, 16 | 1935 | |||
| AF152247.1 | - | 16, 17 | 2101 | |||
| AK173080.1 | - | 12, 13 | 1862 | |||