Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004897.1 | - | 7 | 710 | ||
AK145900.1 | - | 7 | 837 | |||
AK149406.1 | - | 7 | 715 | |||
AK152772.1 | - | 7 | 715 | |||
AK153050.1 | - | 7 | 843 | |||
AK158388.1 | - | 6 | 2623 | |||
AK161189.1 | - | 7 | 689 | |||
AK168078.1 | - | 7 | 715 | |||
BC056644.1 | - | 7 | 699 | 5098645 | ||
U15977.1 | - | 6 | 628 |