Search
Search

Fgf5| Gene Symbol: Fgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277268.1 | NP_001264197.1 | 1, 2 | 588 | ||
| NM_010203.5 | NP_034333.1 | 2, 3 | 692 | |||
| GenBank mRNA | AB016516.1 | - | 1, 2 | 360 | ||
| AK028694.1 | - | 2, 3 | 549 | |||
| AK028894.1 | - | 2, 3 | 694 | |||
| BC071227.1 | - | 2, 3 | 665 | 30443152 | ||
| M30643.1 | - | 2, 3 | 464 | |||