Search
Search

Gnb5| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010313.2 | NP_034443.1 | 12 | 1233 | ||
| NM_138719.5 | NP_619733.1 | 10 | 1209 | |||
| GenBank mRNA | AK050564.1 | - | 10 | 1114 | ||
| AK148175.1 | - | 10 | 1104 | |||
| AK149908.1 | - | 10 | 1026 | |||
| AK162103.1 | - | 2 | 373 | |||
| AK169323.1 | - | 10 | 1142 | |||
| AK206251.1 | - | 1 | 19 | |||
| AK211560.1 | - | 1 | 27 | |||
| AK215403.1 | - | 1 | 27 | |||
| AK216405.1 | - | 1 | 27 | |||
| BC016135.1 | - | 12 | 1167 | 4506201 | ||
| BX513135.1 | - | 1 | 65 | |||
| L34290.1 | - | 10 | 1035 | |||
| U69145.1 | - | 11 | 1131 | |||