Search
Search

Gnb5| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010313.2 | NP_034443.1 | 6, 7 | 593 | ||
| NM_138719.5 | NP_619733.1 | 4, 5 | 569 | |||
| GenBank mRNA | AK018747.1 | - | 2, 3 | 123 | ||
| AK050564.1 | - | 4, 5 | 474 | |||
| AK148175.1 | - | 4, 5 | 464 | |||
| AK149908.1 | - | 4, 5 | 386 | |||
| AK169323.1 | - | 4, 5 | 502 | |||
| BC016135.1 | - | 6, 7 | 527 | 4506201 | ||
| CK621143.1 | - | 6, 7 | 593 | |||
| L34290.1 | - | 4, 5 | 395 | |||
| U69145.1 | - | 5, 6 | 491 | |||