Search
Search

Gnb5| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010313.2 | NP_034443.1 | 12 | 1136 | ||
| NM_138719.5 | NP_619733.1 | 10 | 1112 | |||
| GenBank mRNA | AK050564.1 | - | 10 | 1017 | ||
| AK148175.1 | - | 10 | 1007 | |||
| AK149908.1 | - | 10 | 929 | |||
| AK162103.1 | - | 2 | 276 | |||
| AK169323.1 | - | 10 | 1045 | |||
| BC016135.1 | - | 12 | 1070 | 4506201 | ||
| L34290.1 | - | 10 | 938 | |||
| U69145.1 | - | 11 | 1034 | |||