Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Hoxc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010465.2 | NP_034595.2 | 1 | 220 | ||
GenBank mRNA | AB221685.1 | - | 1 | 176 | ||
BC028482.1 | - | 1 | 220 | |||
BC141061.1 | - | 1 | 199 | 9055673 | ||
BX528590.1 | - | 1 | 221 | |||
X12504.1 | - | 2 | 245 | |||
X16510.1 | - | 181 | ||||
X16511.1 | - | 1 | 303 |