Search
Search

Irf2| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008391.4 | NP_032417.3 | 6 | 671 | ||
| GenBank mRNA | AK036652.1 | - | 6 | 639 | ||
| AK132099.1 | - | 6 | 684 | |||
| AK155025.1 | - | 6 | 656 | |||
| BC006577.1 | - | 6 | 664 | 3586586 | ||
| BC110666.1 | - | 6 | 643 | 4918447 | ||
| J03168.1 | - | 6 | 626 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK163770.1 | - | 3849 | Chr. - - [-] on Build | ||