Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITGAV SIRNA Assays ›
Gene Symbol:
Itgav
Gene Name:
integrin alpha V
Gene Aliases:
1110004F14Rik, 2610028E01Rik, CD51, D430040G12Rik
Chromosome Location:
Chr.2: 83723819 - 83806917 on Build GRCm38
UniGene ID:
Mm.227
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS205549
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Itgav
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008402.3
NP_032428.2
7
996
GenBank mRNA
AK149984.1
-
7
898
BC133682.1
-
7
707
U14135.1
-
7
878
Back To Top
More Information
Panther Classification:
Molecular Function -
integrin
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
blood vessel development
cellular calcium ion homeostasis
inflammatory response
cell adhesion
cell-matrix adhesion
positive regulation of cytosolic calcium ion concentration
integrin-mediated signaling pathway
multicellular organism development
positive regulation of cell proliferation
fertilization
negative regulation of macrophage derived foam cell differentiation
negative regulation of lipid storage
cell growth
cell migration
cell differentiation
positive regulation of cell migration
cell-substrate adhesion
negative regulation of lipid transport
detection of molecule of bacterial origin
positive regulation of osteoblast proliferation
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
regulation of ion transmembrane transport
gonad morphogenesis
endodermal cell differentiation
apolipoprotein A-I-mediated signaling pathway
T cell activation
apoptotic cell clearance
positive regulation of MAPK cascade
endothelial cell migration
regulation of bone resorption
negative regulation of low-density lipoprotein particle receptor biosynthetic process
positive regulation of cell adhesion
viral entry into host cell
focal adhesion assembly
negative regulation of lipoprotein metabolic process
regulation of phagocytosis
negative chemotaxis
release of sequestered calcium ion into cytosol
trophoblast giant cell differentiation
ERK1 and ERK2 cascade
calcium ion transmembrane transport
response to nitric oxide
extrinsic apoptotic signaling pathway in absence of ligand
regulation of apoptotic cell clearance
negative regulation of entry of bacterium into host cell
negative regulation of extrinsic apoptotic signaling pathway
opsonin binding
fibronectin binding
protease binding
protein kinase C binding
receptor binding
voltage-gated calcium channel activity
insulin-like growth factor I binding
peptide binding
metal ion binding
protein heterodimerization activity
extracellular matrix binding
extracellular matrix protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0