Search
Search

Itgb1| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010578.2 | NP_034708.1 | 10 | 1550 | ||
| GenBank mRNA | AK076526.1 | - | 6 | 847 | ||
| AK088016.1 | - | 10 | 1451 | |||
| AK088729.1 | - | 10 | 1297 | |||
| AK135521.1 | - | 9 | 1226 | |||
| AK141253.1 | - | 5 | 477 | |||
| AK159689.1 | - | 10 | 1392 | |||
| AK167682.1 | - | 10 | 1448 | |||
| BC050906.1 | - | 10 | 1423 | 5721348 | ||
| X15202.1 | - | 9 | 1243 | |||
| Y00769.1 | - | 10 | 1338 | |||