Search
Search

Itgb1| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010578.2 | NP_034708.1 | 7 | 1119 | ||
| GenBank mRNA | AK076526.1 | - | 3 | 416 | ||
| AK088016.1 | - | 7 | 1020 | |||
| AK088729.1 | - | 7 | 866 | |||
| AK135521.1 | - | 6 | 795 | |||
| AK141253.1 | - | 2 | 46 | |||
| AK159689.1 | - | 7 | 961 | |||
| AK167682.1 | - | 7 | 1017 | |||
| BC050906.1 | - | 7 | 992 | 5721348 | ||
| X15202.1 | - | 6 | 812 | |||
| Y00769.1 | - | 7 | 907 | |||