Search
Search

Kcnh2| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013569.2 | NP_038597.2 | 5 | 1304 | ||
| GenBank mRNA | AF012868.1 | - | 5 | 1310 | ||
| AY374424.1 | - | 5 | 989 | |||
| BC051016.1 | - | 5 | 1135 | |||
| BQ745654.1 | - | 2 | 198 | |||