Search
Search

Kcnj12| Gene Symbol: Kcnj12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267593.1 | NP_001254522.1 | 2 | 545 | ||
| NM_010603.6 | NP_034733.4 | 3 | 717 | |||
| GenBank mRNA | AB035889.1 | - | 1 | 177 | ||
| AK017299.1 | - | 3 | 718 | |||
| AK032271.1 | - | 2 | 547 | |||
| BC115970.1 | - | 2 | 297 | 40106507 | ||
| BC116677.1 | - | 2 | 297 | 40106509 | ||
| X80417.1 | - | 1 | 282 | |||