Search
Search

Kif9| Gene Symbol: Kif9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163569.1 | NP_001157041.1 | 17, 18 | 2051 | ||
| NM_010628.3 | NP_034758.2 | 17, 18 | 2056 | |||
| GenBank mRNA | AJ132889.1 | - | 17, 18 | 1989 | ||
| AK029506.1 | - | 17, 18 | 1949 | |||
| AK029776.1 | - | 17, 18 | 2021 | |||
| AK077169.1 | - | 9, 10 | 992 | |||