Search
Search

Klc1| Gene Symbol: Klc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025358.2 | NP_001020529.2 | 4 | 708 | ||
| NM_001025359.2 | NP_001020530.2 | 4 | 708 | |||
| NM_001025360.2 | NP_001020531.2 | 4 | 727 | |||
| NM_001025361.2 | NP_001020532.2 | 4 | 708 | |||
| NM_001025362.2 | NP_001020533.2 | 4 | 708 | |||
| NM_001025363.2 | NP_001020534.2 | 4 | 708 | |||
| NM_001081959.1 | NP_001075428.1 | 4 | 727 | |||
| NM_008450.2 | NP_032476.2 | 4 | 727 | |||
| GenBank mRNA | AF055665.1 | - | 4 | 626 | ||
| AK031309.1 | - | 4 | 699 | |||
| AY753300.1 | - | 3 | 532 | |||
| BC055744.1 | - | 4 | 563 | 6415280 | ||
| CF615940.1 | - | 4 | 638 | |||