Search Thermo Fisher Scientific
Gene Symbol: Epcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008532.2 | NP_032558.2 | 10 | 1388 | ||
GenBank mRNA | AK133510.1 | - | 9 | 1070 | ||
AK145752.1 | - | 9 | 964 | |||
AK167182.1 | - | 9 | 1033 | |||
BC005618.1 | - | 9 | 1029 | 3711069 | ||
BC094465.1 | - | 9 | 1054 | 4948935 | ||
M76124.1 | - | 9 | 1037 |