Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134234.1 | NP_598995.1 | 1 | 141 | ||
GenBank mRNA | BC109325.2 | - | 1 | 141 | 40050555 |