Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134252.3 | NP_599013.1 | 14, 15 | 1918 | ||
GenBank mRNA | AF481480.2 | - | 2187 | |||
AY095352.1 | - | 12, 13 | 1752 | |||
BC117934.1 | - | 14, 15 | 1918 | 40097778 |