Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081117.2 | NP_001074586.2 | 12 | 2669 | ||
GenBank mRNA | AK052772.1 | - | 12 | 2589 | ||
AK082099.1 | - | 12 | 2588 | |||
AK143677.1 | - | 12 | 2708 | |||
X82786.1 | - | 12 | 2777 |