Search
Search

Mtnr1a| Gene Symbol: Mtnr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008639.2 | NP_032665.1 | 2 | 341 | ||
| GenBank mRNA | BC116900.1 | - | 2 | 414 | 40126219 | |
| BC119313.1 | - | 2 | 414 | 8734062 | ||
| U52222.1 | - | 2 | 341 | |||
Set Membership: |
|