Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Oit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010959.2 | NP_035089.1 | 7 | 1421 | ||
GenBank mRNA | AF517106.1 | - | 6 | 1240 | ||
AF517107.1 | - | 6 | 1240 | |||
AK040570.1 | - | 7 | 1422 | |||
AK050081.1 | - | 7 | 1388 | |||
AK153440.1 | - | 3 | 663 | |||
AY180915.1 | - | 7 | 1345 |