Search
Search

Pitpna| Gene Symbol: Pitpna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008850.1 | NP_032876.1 | 5, 6 | 595 | ||
| GenBank mRNA | AK144618.1 | - | 6, 7 | 608 | ||
| AK149745.1 | - | 6, 7 | 585 | |||
| AK151496.1 | - | 6, 7 | 584 | |||
| AK152089.1 | - | 6, 7 | 608 | |||
| AK152874.1 | - | 6, 7 | 602 | |||
| AK153569.1 | - | 6, 7 | 608 | |||
| AK165659.1 | - | 6, 7 | 600 | |||
| AK167760.1 | - | 5, 6 | 608 | |||
| AK168187.1 | - | 5, 6 | 565 | |||
| AK168720.1 | - | 6, 7 | 598 | |||
| BC056171.1 | - | 6, 7 | 550 | 5710926 | ||
| S72681.1 | - | 5, 6 | 370 | |||
| U96725.1 | - | 5, 6 | 596 | |||