Search
Search

Pitx1| Gene Symbol: Pitx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011097.2 | NP_035227.1 | 2 | 839 | ||
| GenBank mRNA | AK142455.1 | - | 2 | 667 | ||
| BC012696.1 | - | 2 | 643 | 4192818 | ||
| U54499.1 | - | 2 | 381 | |||
| U71206.1 | - | 2 | 839 | |||