Search
Search

Pitx2| Gene Symbol: Pitx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042502.2 | NP_001035967.1 | 3 | 1317 | ||
| NM_001042504.2 | NP_001035969.1 | 4 | 808 | |||
| NM_001286942.1 | NP_001273871.1 | 5 | 907 | |||
| NM_001287048.1 | NP_001273977.1 | 3 | 1317 | |||
| NM_011098.4 | NP_035228.2 | 5 | 946 | |||
| GenBank mRNA | AB006320.1 | - | 3 | 870 | ||
| AB006321.1 | - | 5 | 906 | |||
| AF048723.1 | - | 4 | 745 | |||
| AF048724.1 | - | 3 | 1278 | |||
| AF201091.1 | - | 3 | 644 | |||
| AM940438.1 | - | 4 | 612 | |||
| AM940439.1 | - | 3 | 570 | |||
| BC075660.1 | - | 3 | 982 | 30606653 | ||
| U70132.1 | - | 2 | 467 | |||
| U80010.1 | - | 5 | 881 | |||
| U80011.1 | - | 4 | 761 | |||
| U80036.1 | - | 4 | 779 | |||