Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pkp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019645.2 | NP_062619.1 | 1 | 148 | ||
GenBank mRNA | AK036856.1 | - | 1 | 332 | ||
AK079429.1 | - | 1 | 332 | |||
BC139041.1 | - | 1 | 243 | 8862063 | ||
BC139042.1 | - | 1 | 244 | 8862064 | ||
Y07941.1 | - | 1 | 148 |