Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKDC SIRNA Assays ›
          Gene Symbol:
          Prkdc
          Gene Name:
          protein kinase, DNA activated, catalytic polypeptide
          Gene Aliases:
          AI326420, AU019811, DNA-PKcs, DNAPDcs, DNAPK, DNPK1, DOXNPH, HYRC1, XRCC7, dxnph, p460, scid, slip
          Chromosome Location:
          Chr.16: 15637444 - 15842252 on Build GRCm38
          UniGene ID:
          Mm.71
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS207961
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Prkdc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011159.2 NP_035289.2 70 9848
          GenBank mRNA AB007544.1 - 70 9825
          AB011543.1 - 70 9837
          D87521.1 - 70 9825

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          double-strand break repair via homologous recombination
          somitogenesis
          negative regulation of protein phosphorylation
          B cell lineage commitment
          pro-B cell differentiation
          T cell lineage commitment
          immunoglobulin production
          negative regulation of immunoglobulin production
          positive regulation of immune system process
          DNA repair
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          protein phosphorylation
          cellular response to DNA damage stimulus
          brain development
          heart development
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to ionizing radiation
          response to gamma radiation
          phosphorylation
          peptidyl-serine phosphorylation
          lymphocyte differentiation
          protein destabilization
          cellular response to insulin stimulus
          T cell differentiation in thymus
          V(D)J recombination
          immunoglobulin V(D)J recombination
          T cell receptor V(D)J recombination
          ectopic germ cell programmed cell death
          regulation of circadian rhythm
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          rhythmic process
          spleen development
          thymus development
          positive regulation of developmental growth
          signal transduction involved in mitotic G1 DNA damage checkpoint
          negative regulation of cellular senescence
          negative regulation of response to gamma radiation
          nucleotide binding
          DNA binding
          double-stranded DNA binding
          protein kinase activity
          protein serine/threonine kinase activity
          DNA-dependent protein kinase activity
          protein binding
          ATP binding
          transcription factor binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phosphotransferase activity, alcohol group as acceptor
          enzyme binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0