Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dnajc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008929.3 | NP_032955.2 | 5 | 590 | ||
GenBank mRNA | AK146097.1 | - | 5 | 554 | ||
AK148270.1 | - | 5 | 547 | |||
BC013766.1 | - | 5 | 535 | 2646147 | ||
U28423.1 | - | 5 | 533 |