Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008941.3 | NP_032967.1 | 19 | 2123 | ||
NM_178855.4 | NP_849186.2 | 18 | 2078 | |||
GenBank mRNA | AK033582.1 | - | 18 | 2080 | ||
AK038356.1 | - | 10 | 1072 | |||
BC117917.1 | - | 19 | 2122 | 40097367 | ||
BC117918.1 | - | 19 | 2122 | 40097370 | ||
U73378.1 | - | 19 | 2118 |