Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008987.3 | NP_033013.3 | 3 | 1306 | ||
GenBank mRNA | BC022176.1 | - | 3 | 1306 | 4012053 | |
X83601.1 | - | 3 | 1230 |