Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Rasal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281999.1 | NP_001268928.1 | 5 | 618 | ||
NM_013832.4 | NP_038860.2 | 6 | 589 | |||
GenBank mRNA | AF086714.1 | - | 5 | 461 | ||
AK156050.1 | - | 5 | 582 | |||
BC005418.1 | - | 6 | 603 | 3488024 |