Search
Search

Stam| Gene Symbol: Stam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011484.2 | NP_035614.1 | 7 | 791 | ||
| GenBank mRNA | AK035924.1 | - | 7 | 714 | ||
| AK144987.1 | - | 7 | 808 | |||
| AK147832.1 | - | 7 | 701 | |||
| AK157767.1 | - | 7 | 738 | |||
| AK162431.1 | - | 7 | 738 | |||
| AK163720.1 | - | 7 | 739 | |||
| AK163922.1 | - | 7 | 728 | |||
| AK169170.1 | - | 7 | 798 | |||
| BC044666.1 | - | 7 | 791 | 4950569 | ||
| BC055326.1 | - | 7 | 737 | 6416552 | ||
| U43900.1 | - | 7 | 648 | |||