Search
Search

Ctnna3| Gene Symbol: Ctnna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164376.1 | NP_001157848.1 | 5, 6 | 732 | ||
| NM_177612.3 | NP_808280.2 | 5, 6 | 660 | |||
| GenBank mRNA | AF344871.1 | - | 5, 6 | 732 | ||
| AK030166.1 | - | 5, 6 | 661 | |||
| AK142745.1 | - | 5, 6 | 733 | |||
| BC112409.1 | - | 5, 6 | 634 | 40060582 | ||
| DQ380429.1 | - | 5, 6 | 660 | |||