Search
Search

Rab3ip| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003950.2 | NP_001003950.1 | 5, 6 | 946 | ||
| GenBank mRNA | AK152122.1 | - | 6, 7 | 1093 | ||
| AK206753.1 | - | 1, 2 | 166 | |||
| BC080289.1 | - | 5, 6 | 946 | 6816077 | ||
| S79539.1 | - | 3, 4 | 1026 | |||