Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NLGN2 SIRNA Assays ›
Gene Symbol:
Nlgn2
Gene Name:
neuroligin 2
Gene Aliases:
NL2
Chromosome Location:
Chr.11: 69823123 - 69838377 on Build GRCm38
UniGene ID:
Mm.151293
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS211270
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nlgn2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_198862.2
NP_942562.2
5
1302
GenBank mRNA
AK136883.1
-
6
1868
AK173159.1
-
5
1302
BE952199.1
-
5
506
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_020795, AB037787, AF376802.
This siRNA also targets the following transcripts in Rat: NM_053992, U41662.
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
thigmotaxis
regulation of respiratory gaseous exchange by neurological system process
cell adhesion
neuron cell-cell adhesion
synapse assembly
jump response
positive regulation of cell proliferation
sensory perception of pain
positive regulation of insulin secretion
positive regulation of synaptic transmission, GABAergic
social behavior
protein localization to synapse
locomotory exploration behavior
modulation of synaptic transmission
synapse organization
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
positive regulation of dendritic spine development
terminal button organization
postsynaptic membrane assembly
presynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
postsynaptic density protein 95 clustering
positive regulation of inhibitory postsynaptic potential
insulin metabolic process
positive regulation of protein localization to synapse
positive regulation of t-SNARE clustering
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
positive regulation of synaptic vesicle clustering
receptor activity
protein binding
neurexin family protein binding
cell adhesion molecule binding
carboxylic ester hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0