siRNA Details
Gene Symbol: Nle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145431.2 | NP_663406.2 | 7 | 725 | ||
GenBank mRNA | AK141230.1 | - | 7 | 700 | ||
AK170853.1 | - | 7 | 698 | |||
BC018399.1 | - | 7 | 716 | 3491925 | ||
BC035421.1 | - | 7 | 716 |
