Search
Search

Ahsa1| Gene Symbol: Ahsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146036.1 | NP_666148.1 | 8 | 945 | ||
| GenBank mRNA | AK076069.1 | - | 8 | 955 | ||
| AK165289.1 | - | 8 | 964 | |||
| AK166644.1 | - | 8 | 918 | |||
| AK182129.1 | - | 2 | 48 | |||
| AK191114.1 | - | 2 | 54 | |||
| BC023857.1 | - | 8 | 912 | 5346745 | ||
| BC025552.1 | - | 8 | 945 | 5320590 | ||
| BC036967.1 | - | 5 | 1513 | |||