Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Amph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289546.1 | NP_001276475.1 | 6 | 503 | ||
NM_175007.2 | NP_778172.1 | 6 | 503 | |||
GenBank mRNA | AK047144.1 | - | 6 | 457 | ||
AK132168.1 | - | 6 | 504 | |||
BC054718.1 | - | 6 | 483 | 5686478 |