Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rnf144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170643.1 | NP_001164114.1 | 5 | 612 | ||
NM_146042.4 | NP_666154.3 | 5 | 648 | |||
GenBank mRNA | AK040939.1 | - | 5 | 604 | ||
AK052846.1 | - | 5 | 613 | |||
AK053529.1 | - | 5 | 648 | |||
BB661852.1 | - | 5 | 613 | |||
BC025007.1 | - | 5 | 592 | 5050848 |