Search
Search

Tmpo| Gene Symbol: Tmpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080129.2 | NP_001073598.1 | 2 | 670 | ||
| NM_001080130.2 | NP_001073599.1 | 2 | 670 | |||
| NM_001080131.2 | NP_001073600.1 | 2 | 670 | |||
| NM_001080132.2 | NP_001073601.1 | 2 | 670 | |||
| NM_001283048.1 | NP_001269977.1 | 2 | 353 | |||
| NM_011605.3 | NP_035735.2 | 2 | 670 | |||
| GenBank mRNA | AK011988.1 | - | 2 | 163 | ||
| AK011992.1 | - | 2 | 163 | |||
| AK134216.1 | - | 2 | 163 | |||
| AK141845.1 | - | 2 | 620 | |||
| AK144642.1 | - | 2 | 630 | |||
| AK146903.1 | - | 2 | 637 | |||
| AK150305.1 | - | 2 | 618 | |||
| AK150515.1 | - | 2 | 605 | |||
| AK151936.1 | - | 2 | 622 | |||
| AK152737.1 | - | 2 | 622 | |||
| AK153281.1 | - | 2 | 605 | |||
| AK156206.1 | - | 2 | 620 | |||
| AK159161.1 | - | 2 | 266 | |||
| AK165282.1 | - | 2 | 620 | |||
| AK165851.1 | - | 2 | 627 | |||
| AK192607.1 | - | 2 | 110 | |||
| BC141062.1 | - | 2 | 388 | 9055674 | ||
| BE852115.1 | - | 2 | 111 | |||
| U39073.1 | - | 2 | 573 | |||
| U39074.1 | - | 2 | 601 | |||
| U39075.1 | - | 2 | 601 | |||
| U39076.1 | - | 2 | 601 | |||
| U39077.1 | - | 2 | 601 | |||
| U39078.1 | - | 2 | 584 | |||