Search
Search

Top3b| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011624.2 | NP_035754.1 | 3 | 429 | ||
| GenBank mRNA | AB013603.1 | - | 2 | 242 | ||
| AB045324.1 | - | 3 | 503 | |||
| AK041159.1 | - | 3 | 339 | |||
| AK079765.1 | - | 3 | 356 | |||
| AK144709.1 | - | 4 | 415 | |||
| AK146475.1 | - | 3 | 451 | |||
| BC006614.1 | - | 3 | 332 | |||
| BC031723.1 | - | 3 | 429 | 5003002 | ||