Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vps45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013841.3 | NP_038869.1 | 12 | 1963 | ||
GenBank mRNA | AK146190.1 | - | 12 | 1365 | ||
AK168101.1 | - | 12 | 1365 | |||
BC012691.1 | - | 12 | 1339 | 4038142 | ||
BC058528.1 | - | 12 | 1355 | 5718352 | ||
U66865.1 | - | 12 | 1978 |