This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
Wnt2
- Gene Name:
-
wingless-type MMTV integration site family, member 2
- Gene Aliases:
-
2610510E18Rik, Int1l1, Irp, Mirp, Wnt-2, Wnt2a
- Chromosome Location:
-
Chr.6: 17985474 - 18032751 on Build GRCm38
- UniGene ID:
-
Mm.33653
- Species:
- Mouse
- Species Specific ID (Flybase ID):
-
-
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System