Search
Search

Dctn6| Gene Symbol: Dctn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293757.1 | NP_001280686.1 | 6 | 698 | ||
| NM_001293758.1 | NP_001280687.1 | 5 | 531 | |||
| NM_001293759.1 | NP_001280688.1 | 6 | 768 | |||
| NM_011722.4 | NP_035852.1 | 5 | 585 | |||
| GenBank mRNA | AF124788.1 | - | 5 | 316 | ||
| AF190796.1 | - | 5 | 326 | |||
| AK003618.1 | - | 5 | 325 | |||
| AK088312.1 | - | 5 | 332 | |||
| BC029249.1 | - | 5 | 326 | 4486579 | ||
| CV560309.1 | - | 5 | 482 | |||
| DV042569.1 | - | 5 | 540 | |||
| DV062065.1 | - | 6 | 490 | |||