Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081073.1 | NP_001074542.1 | 7, 8 | 1100 | ||
GenBank mRNA | AK035879.1 | - | 6, 7 | 880 | ||
BC119216.1 | - | 7, 8 | 981 | 8733965 | ||
BC119218.1 | - | 7, 8 | 981 | 8733967 |