Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172462.4 | NP_766050.1 | 3 | 1650 | ||
GenBank mRNA | AK032872.1 | - | 3 | 1665 | ||
AK085077.1 | - | 2 | 1636 | |||
BC058553.1 | - | 3 | 1648 | |||
BC094590.1 | - | 2 | 1635 | 30616053 | ||
BC139312.1 | - | 2 | 1630 | 8862334 | ||
BC139315.1 | - | 2 | 1630 | 8862337 |