Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GSN SIRNA Assays ›
          Gene Symbol:
          Gsn
          Gene Name:
          gelsolin
          Gene Aliases:
          ADF
          Chromosome Location:
          Chr.2: 35256359 - 35307902 on Build GRCm38
          UniGene ID:
          Mm.21109
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS213462
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gsn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206367.1 NP_001193296.1 18 2359
          NM_001206368.1 NP_001193297.1 17 2275
          NM_001206369.1 NP_001193298.1 17 2320
          NM_146120.4 NP_666232.2 17 2382
          GenBank mRNA AK076156.1 - 17 2322
          AK089934.1 - 18 2314
          AK135632.1 - 17 2093
          AK143664.1 - 17 2349
          AK146969.1 - 18 2276
          AK148957.1 - 18 2339
          AK150359.1 - 18 2282
          AK151684.1 - 18 2301
          AK153029.1 - 18 2279
          AK166831.1 - 17 2295
          AK166932.1 - 18 2273
          AK167918.1 - 18 2289
          AK168461.1 - 18 2292
          AK168532.1 - 18 2293
          AK168707.1 - 17 2228
          AK168725.1 - 18 2276
          AK168829.1 - 17 2328
          AK169041.1 - 18 2279
          AK169670.1 - 18 2304
          BC023143.1 - 17 2320 3711602
          BC060377.1 - 18 2307 4983664
          BY655071.1 - 1 124
          J04953.1 - 16 2169

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagocytosis, engulfment
          apoptotic process
          actin polymerization or depolymerization
          positive regulation of gene expression
          striated muscle atrophy
          vesicle-mediated transport
          cell projection organization
          actin filament polymerization
          regulation of cell adhesion
          protein destabilization
          sequestering of actin monomers
          actin nucleation
          negative regulation of viral entry into host cell
          phosphatidylinositol-mediated signaling
          actin filament severing
          barbed-end actin filament capping
          positive regulation of actin nucleation
          actin filament capping
          cilium morphogenesis
          cellular response to cadmium ion
          regulation of podosome assembly
          actin filament reorganization
          renal protein absorption
          hepatocyte apoptotic process
          positive regulation of keratinocyte apoptotic process
          regulation of wound healing, spreading of epidermal cells
          regulation of establishment of T cell polarity
          regulation of plasma membrane raft polarization
          regulation of receptor clustering
          positive regulation of protein processing in phagocytic vesicle
          amyloid fibril formation
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          actin binding
          calcium ion binding
          protein binding
          protein domain specific binding
          myosin II binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0