Search
Search

Galnt11| Gene Symbol: Galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144908.3 | NP_659157.1 | 7 | 1208 | ||
| GenBank mRNA | AF495528.1 | - | 6 | 1080 | ||
| AK088014.1 | - | 7 | 1197 | |||
| AK161524.1 | - | 7 | 1118 | |||
| BC011428.1 | - | 7 | 1184 | 2631696 | ||
| BC021504.1 | - | 7 | 1209 | 5346678 | ||
| BC031150.1 | - | 6 | 1106 | |||
| BC034184.1 | - | 6 | 1108 | 4974216 | ||
| BC034185.1 | - | 6 | 1160 | 4974695 | ||
| BC036143.1 | - | 6 | 1109 | 4974645 | ||
| BC036145.1 | - | 6 | 1102 | 4975474 | ||
| Y12435.1 | - | 6 | 1031 | |||